Tin(IV) isopropoxide 10% (w/v) in 2-propanol/toluene , 98% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
36563.AA
36563.AE
36563.AAEA
41.2
EUR
36563.AA
36563.AE
Tin(IV) isopropoxide 10% (w/v) in 2-propanol/toluene , 98% (metals basis)
Tin(IV) isopropoxide
Purity: 98%
CAS No.: 1184-61-8
Molecular Formula: Sn[OCH(CH3)2]4
Formula Weight: 355.06
Physical Form: Liquid
Sensitivity: Moisture Sensitive
UN No.: UN1993
MDL No.: MFCD00145408
Formule:
C₁₂H₂₈O₄Sn MW: 355.062 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00145408 CAS nummer: 1184-61-8 UN: 1993 ADR: 3 II |