Niobium isopropoxide 10% (w/v) in isopropanol/hexane (50:50) , 99% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
36572.AC
36572.AE
36572.ACEA
178
EUR
36572.AC
36572.AE
Niobium isopropoxide 10% (w/v) in isopropanol/hexane (50:50) , 99% (metals basis)
Niobium isopropoxide
Purity: 99%
CAS No.: 18368-80-4
Molecular Formula: C15H35NbO5
Formula Weight: 388.35
Physical Form: Liquid
Sensitivity: Moisture Sensitive
UN No.: UN1993
MDL No.: MFCD00144915
Formule:
C₁₅H₃₅NbO₅ MW: 388.346 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00144915 CAS nummer: 18368-80-4 UN: 1993 ADR: 3 II |