Aluminum calcium isopropoxide 10% (w/v) in isopropanol (IPA) , 99.8% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
36594.AC
36594.AE
36594.ACEA
89.5
EUR
36594.AC
36594.AE
Aluminum calcium isopropoxide 10% (w/v) in isopropanol (IPA) , 99.8% (metals basis)
Aluminum calcium isopropoxide
Purity: 99.8%
CAS No.: 23275-27-6
Molecular Formula: Ca[Al(OCH(CH3)2)4]2
Formula Weight: 566.30
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Odour: Alcohol-like
Boiling Point: 230-240°/4.0mm
UN No.: UN1219
MDL No.: MFCD00143586
Formule:
Ca[Al(OCH(CH₃)₂)₄]₂ Kookpunt: 230…240 °C (4,0 mmHg) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00143586 CAS nummer: 23275-27-6 UN: 1219 ADR: 3,II |