Semiquantitative Standard 2, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36770.AE 36770.AP
36770.AEEA 546 EUR
36770.AE 36770.AP
Semiquantitative Standard 2, Specpure®
Standaarden Kationische standaarden Standaarden met meerdere elementen
Molecular Formula: Matrix: 40% Aqua Regia/tr. HF
Physical Form: Liquid
UN No.: UN3093
MDL No.: MFCD02091743