Palladium 3% on activated carbon wetted with ca. 50% water, powder hydrogenation catalyst, reduced
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
38296.06 38296.14 38296.22
38296.06EA 57.8 EUR
38296.06 38296.14 38296.22
Palladium 3% on activated carbon wetted with ca. 50% water, powder hydrogenation catalyst, reduced
Palladium
CAS No.: 7440-05-3 Physical Form: 900-1100m2/g Odour: Odourless MDL No.: MFCD03457879
Formule: Pd
MW: 106.420 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011167
CAS nummer: 7440-05-3