Palladium 10% on activated carbon powder wetted with ca. 50% water for use as a catalyst, reduced
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
38304.06 38304.14 38304.22
38304.06EA 165 EUR
38304.06 38304.14 38304.22
Palladium 10% on activated carbon powder wetted with ca. 50% water for use as a catalyst, reduced
Palladium
Physical Form: 900-1100 m²/g
Formule: Pd
MW: 106.420 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011167
CAS nummer: 7440-05-3