Platinum 10% on activated carbon wetted with ca. 50% water, powder, unreduced
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Platinium
38326.06 38326.14 38326.22
38326.06EA 139 EUR
38326.06 38326.14 38326.22
Platinum 10% on activated carbon wetted with ca. 50% water, powder, unreduced
Platinum
CAS No.: 7440-06-4 Physical Form: 900-1100 m²/g MDL No.: MFCD00011179
Formule: Pt
MW: 195.08 g/mol
Kookpunt: 3827
Smeltpunt: 1769
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011179
CAS nummer: 7440-06-4
Merck Index: 12,07689