Iridium 0,5% on activated carbon wetted with ca. 50% water, powder, reduced
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
38327.14 38327.22 38327.06
38327.14EA 274 EUR
38327.14 38327.22 38327.06
Iridium 0,5% on activated carbon wetted with ca. 50% water, powder, reduced
Iridium
MDL No.: MFCD00011062
Formule: Ir
MW: 192.22 g/mol
Kookpunt: 5300
Smeltpunt: 2450
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011062
CAS nummer: 7439-88-5