Multi-element QC-19 check standard solution, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
39146.AE 39146.AP
39146.AEEA 431 EUR
39146.AE 39146.AP
Multi-element QC-19 check standard solution, Specpure®
Standaarden Kationische standaarden Standaarden met meerdere elementen
Molecular Formula: Matrix: 5% HNO3/tr. F-/tr. tartaric acid
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00210655