Lead(II) titanium(IV) isopropoxide 5% (w/v) in isopropanol (IPA) , 99.9% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
39153.AC
39153.AE
39153.ACEA
109
EUR
39153.AC
39153.AE
Lead(II) titanium(IV) isopropoxide 5% (w/v) in isopropanol (IPA) , 99.9% (metals basis)
Lead(II) titanium(IV) isopropoxide
Purity: 99.9%
Molecular Formula: PbTi(OC3H7)6
Formula Weight: 609.29
Flash Point: 12°(54°F)
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown
Odour: Alcohol-like
UN No.: UN1219
MDL No.: MFCD00210621
Formule:
PbTi(OC₃H₇)₆ MW: 609.29 g/mol Vlampunt: 12 °C (54 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00210621 UN: 1219 ADR: 3 II |