Iodotrimethylplatinum(IV)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
39286.02
39286.03
39286.02EA
253
EUR
39286.02
39286.03
Iodotrimethylplatinum(IV)
Iodotrimethylplatinum(IV)
CAS No.: 14364-93-3
Molecular Formula: C3H9IPt
Formula Weight: 367.09
Physical Form: Crystalline
Sensitivity: Air and Light Sensitive
Appearance: White to pale orange
Odour: Odourless
MDL No.: MFCD00013702
Formule:
C₃H₉IPt Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00013702 CAS nummer: 14364-93-3 |