Lithium isobutyrate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
39372.14
39372.36
39372.14EA
39.9
EUR
39372.14
39372.36
Lithium isobutyrate ≥98%
Lithium isobutyrate
Purity: 98+%
CAS No.: 25179-23-1
Molecular Formula: LiO2CCH(CH3)2
Formula Weight: 94.03
Physical Form: Powder/ lump
Sensitivity: Moisture Sensitive
MDL No.: MFCD00210622
Formule:
LiO₂CCH(CH₃)₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00210622 CAS nummer: 25179-23-1 |