Magnesium salicylate tetrahydrate, tech.
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
39379.22
39379.36
39379.A4
39379.22EA
40.8
EUR
39379.22
39379.36
39379.A4
Magnesium salicylate tetrahydrate, tech.
Magnesium salicylate tetrahydrate
CAS No.: 18917-95-8
Molecular Formula: C14H10MgO6.4H2O
Formula Weight: 370.60 (298.60anhy)
Physical Form: Powder
Sensitivity: Air Sensitive
Odour: Odourless
Solubility: Soluble in water and alcohol
MDL No.: MFCD00045815
Formule:
C₁₄H₁₀MgO₆·4H₂O MW: 370.60 g/mol (298.60 anhy) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00045815 CAS nummer: 18917-95-8 EINECS: 242-669-3 Merck Index: 14,5672 |