Tungstosilicic acid hydrate, Reagent Grade
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
CAS No.: 12027-43-9
Molecular Formula: H4SiO4.12WO3.xH2O
Formula Weight: 2878.28(anhy)
Physical Form: Crystalline
Solubility: Soluble in water
MDL No.: MFCD00036271
Applications: Catalyst for organic synthesis, minerals separation, reagent for alkaloids
Formule:
H₄Si(W₃O₁₀)₄.xH₂O Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00036271 CAS nummer: 12027-43-9 Merck Index: 14,8502 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...