Cerium(IV)-2-methoxyethoxide 18 - 20% (w/w) in 2-methoxyethanol
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
40471.06
40471.14
40471.22
40471.06EA
89.9
EUR
40471.06
40471.14
40471.22
Cerium(IV)-2-methoxyethoxide 18 - 20% (w/w) in 2-methoxyethanol
Cerium(IV)-2-methoxyethoxide
CAS No.: 876107-33-4
Molecular Formula: C12H28CeO8
Formula Weight: 440.47
Physical Form: Liquid
Density: 1.02
Sensitivity: Moisture Sensitive
Appearance: Yellow
UN No.: UN1188
MDL No.: MFCD00236530
Formule:
C₁₂H₂₈CeO₈ Dichtheid: 1,02 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00236530 CAS nummer: 876107-33-4 UN: 1188 ADR: 3,III |