Aluminum Magnesium (alloy 5056), gauze woven from wire 30 mesh woven from 0.23mm (0.009in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40906.FL
40906.GT
40906.FLEA
94.8
EUR
40906.FL
40906.GT
Aluminum Magnesium (alloy 5056), gauze woven from wire 30 mesh woven from 0.23mm (0.009in)
Aluminum magnesium (alloy 5056)
Molecular Formula: Al:Mg; 95:5 wt%
Physical Form: Wire Cloth
Odour: Odourless
MDL No.: MFCD00214039