Hastelloy C® alloy, Draadgaas 20 mesh woven from 0.23mm (0.009in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
40938.FL
40938.GT
40938.HB
40938.FLEA
38.2
EUR
40938.FL
40938.GT
40938.HB
Hastelloy C® alloy, Draadgaas 20 mesh woven from 0.23mm (0.009in)
Hastelloy C® alloy
Physical Form: Wire Cloth
MDL No.: MFCD00801065
Applications: High strength, corrosion resistant alloy