Hastelloy C® alloy, gauze 100 mesh woven from wire 0.102 mm (0.004in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
40940.FL
40940.GT
40940.HB
40940.FLEA
50.2
EUR
40940.FL
40940.GT
40940.HB
Hastelloy C® alloy, gauze 100 mesh woven from wire 0.102 mm (0.004in)
Hastelloy C® alloy
Physical Form: Wire Cloth
MDL No.: MFCD00801065