Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze_wire 20 mesh woven from 0.102mm (0.004in) Type 316
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40943.FL
40943.GT
40943.HB
40943.FLEA
40.1
EUR
40943.FL
40943.GT
40943.HB
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze_wire 20 mesh woven from 0.102mm (0.004in) Type 316
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
Physical Form: Wire Cloth
MDL No.: MFCD00148499