Lead tin, solder alloy Pb:Sn - 50:50 %, Ø 3.18 mm (0.125 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
41023.29
41023.TB
41023.29EA
89.6
EUR
41023.29
41023.TB
Lead tin, solder alloy Pb:Sn - 50:50 %, Ø 3.18 mm (0.125 in)
Lead tin, solder alloy
Molecular Formula: Pb:Sn; 50:50 wt%
MDL No.: MFCD00798549
Applications: For use with copper, copper alloys, lead, steel, and nickel alloys. Recommended for light electrical wiring