Lead tin, solder alloy, with rosin core Pb:Sn - 50:50 %, Ø 1.6 mm (0.06 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
41024.29
41024.TB
41024.29EA
113
EUR
41024.29
41024.TB
Lead tin, solder alloy, with rosin core Pb:Sn - 50:50 %, Ø 1.6 mm (0.06 in)
Lead tin, solder alloy, with rosin core
Molecular Formula: Pb:Sn; 50:50 wt%
MDL No.: MFCD00798549