Cobalt(II)-2-methoxyethoxide 5% (w/v) in 2-methoxyethanol , 99% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
41237.AC
41237.AE
41237.ACEA
130
EUR
41237.AC
41237.AE
Cobalt(II)-2-methoxyethoxide 5% (w/v) in 2-methoxyethanol , 99% (metals basis)
Cobalt(II)-2-methoxyethoxide
Purity: 99%
CAS No.: 142600-61-1
Molecular Formula: C6H14CoO4
Formula Weight: 209.11
Flash Point: 40°(104°F)
Physical Form: Liquid
Density: 0.96
Sensitivity: Moisture Sensitive
Appearance: Dark brown
Boiling Point: 124°
UN No.: UN1188
MDL No.: MFCD00798554
Formule:
C₆H₁₄CoO₄ Kookpunt: 124 °C Dichtheid: 0,96 Vlampunt: 40 °C (104 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00798554 CAS nummer: 142600-61-1 UN: 1188 ADR: 3,III |