Polyvinyl alcohol, M.W. ∼ 88,000-97,000 98-99%, high molecular weight, hydrolysed
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41243.14
41243.22
41243.36
41243.14EA
24
EUR
41243.14
41243.22
41243.36
Polyvinyl alcohol, M.W. ∼ 88,000-97,000 98-99%, high molecular weight, hydrolysed
Polyvinyl alcohol
Purity: 98-99%
CAS No.: 9002-89-5
Molecular Formula: [-CH2CH(OH)-]n
Physical Form: Solid
Odour: Odourless
MDL No.: MFCD00081922
Formule:
[-CH₂CH(OH)-]n Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00081922 CAS nummer: 9002-89-5 |