Tin(IV) isopropoxide isopropanol adduct ≥98% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
41269.03
41269.06
41269.03EA
60.3
EUR
41269.03
41269.06
Tin(IV) isopropoxide isopropanol adduct ≥98% (metals basis)
Tin(IV) isopropoxide isopropanol adduct
Purity: 98%
CAS No.: 132951-93-0
Molecular Formula: Sn[OCH(CH3)2]4.C3H7OH
Formula Weight: 414.15
Physical Form: Wet solid
Sensitivity: Moisture Sensitive
Melting Point: ca 130° dec.
Boiling Point: 131°/1.6mm
MDL No.: MFCD00145408
Formule:
Sn[OCH(CH₃)₂]₄·C₃H₇OH Kookpunt: 131 °C (1,6 mmHg) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00145408 CAS nummer: 132951-93-0 |