Multi-element QC-21 check standard solution, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41328.AE 41328.AP
41328.AEEA 433 EUR
41328.AE 41328.AP
Multi-element QC-21 check standard solution, Specpure®
Standaarden Kationische standaarden Standaarden met meerdere elementen
Multi-Element QC-21 Check Standard Solution is used as a certified reference material or quality control check standard for inductively coupled plasma optical emission spectroscopy (ICP-OES). It is also used in inductively coupled plasma mass spectrometry (ICP-MS), and flame or furnace atomic absorption spectroscopy.
Molecular Formula: Matrix: 5% HNO₃/tr. F-/tr. tartaric acid
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00803610