Vinylene carbonate ≥97% stabilised
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Stabilisatie:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Stab. with BHT
Purity: 97+%
CAS No.: 872-36-6
Molecular Formula: C3H2O3
Formula Weight: 86.05
Storage Temperature: 2°C to 8°C
Flash Point: 72°(163°F)
Physical Form: Liquid
Density: 1.355
Refractive Index: 1.4210
Sensitivity: Moisture Sensitive
Appearance: Colourless to pale yellow
Melting Point: 19-22°
Boiling Point: 162-163°
MDL No.: MFCD00005380
Formule:
C₃H₂O₃ Kookpunt: 162…163 °C Smeltpunt: 19…22 °C Dichtheid: 1,355 Vlampunt: 72 °C (163 °F) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00005380 CAS nummer: 872-36-6 EINECS: 212-825-5 |