Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.1 mm (0.004 in) Type 304
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
41583.FF
41583.FI
41583.GH
41583.FFEA
35.7
EUR
41583.FF
41583.FI
41583.GH
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.1 mm (0.004 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Odour: Odourless
MDL No.: MFCD00148499