Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.5 mm (0.02 in) Type 304
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41585.FF
41585.FI
41585.GH
41585.FFEA
35.2
EUR
41585.FF
41585.FI
41585.GH
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.5 mm (0.02 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Odour: Odourless
MDL No.: MFCD00148499