Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire, twilled weave 400 mesh woven from 0.028mm (0.0011in) Type 304
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
42012.CF
42012.VA
42012.CFEA
258
EUR
42012.CF
42012.VA
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire, twilled weave 400 mesh woven from 0.028mm (0.0011in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Physical Form: Wire Cloth
Odour: Odourless
MDL No.: MFCD00148499