Hexavalent chromium, standard solution, Cr+6 1000 µg/ml, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42234.AE 42234.AP
42234.AEEA 124 EUR
42234.AE 42234.AP
Hexavalent chromium, standard solution, Cr+6 1000 µg/ml, Specpure®
Standaarden Kationische standaarden Chroom standaarden
Molecular Formula: (NH₄)₂Cr₂O₇ in H₂O
Physical Form: Liquid
Appearance: Yellow
Odour: Odourless
MDL No.: MFCD00717544