Iodine monochloride ∼0.22 N in glacial acetic acid
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Iodochloride
42647.AP 42647.K2
42647.APEA 86.9 EUR
42647.AP 42647.K2
Iodine monochloride ∼0.22 N in glacial acetic acid
Iodine monochloride
CAS No.: 7790-99-0 Molecular Formula: ClI Formula Weight: 162.36 Physical Form: Liquid Sensitivity: Moisture and Light Sensitive UN No.: UN2920 MDL No.: MFCD00011354
Applications: For determination of iodine absorption number of fats
Formule: ClI
MW: 162.36 g/mol
Kookpunt: 97.3
Smeltpunt: 26
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011354
CAS nummer: 7790-99-0
EINECS: 232-236-7
UN: 2920
ADR: 8 II
Merck Index: 13,05039