Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, annealed, Ø 0.25 mm (0.01 in) Type 304
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
43300.BY
43300.BU
43300.CC
43300.BYEA
45.2
EUR
43300.BY
43300.BU
43300.CC
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, annealed, Ø 0.25 mm (0.01 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Physical Form: Temper: soft
Odour: Odourless
MDL No.: MFCD00148499