Lithium tri-tert-butoxyaluminum hydride 1 M in THF
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Lithium tri-tert-butoxyaluminohydride
Packaged under argon in resealable ChemSeal™ bottles.
43324.AB
43324.AD
43324.ABEA
44.7
EUR
43324.AB
43324.AD
Lithium tri-tert-butoxyaluminum hydride 1 M in THF
Lithium tri-tert-butoxyaluminum hydride
Packaged under Argon in resealable ChemSeal™ bottles
CAS No.: 17476-04-9
Molecular Formula: C12H28AlLiO3
Formula Weight: 254.28
Flash Point: -17°(1°F)
Physical Form: Liquid
Density: 0.904
Sensitivity: Moisture Sensitive
Appearance: Colourless
UN No.: UN3399
MDL No.: MFCD00011532
Formule:
C₁₂H₂₈AlLiO₃ Dichtheid: 0,904 Vlampunt: –17 °C (1 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011532 CAS nummer: 17476-04-9 UN: 3399 ADR: 4.3,I |