Nickel iron copper molybdenum (77:14:5:4 wt%), foil, annealed, Thickness: 0.125 mm (0.005 in) (Mumetal™)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
43399.GT
43399.FL
43399.GTEA
343
EUR
43399.GT
43399.FL
Nickel iron copper molybdenum (77:14:5:4 wt%), foil, annealed, Thickness: 0.125 mm (0.005 in) (Mumetal™)
Nickel iron copper molybdenum
Applications: Magnetic shielding alloy
Molecular Formula: Ni:Fe:Cu:Mo; 77:14:5:4 wt%
MDL No.: MFCD02684505
Applications: Magnetic shielding alloy