Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), powder -10+20 mesh Type 316-L
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
43458.A3
43458.36
43458.A3EA
239
EUR
43458.A3
43458.36
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), powder -10+20 mesh Type 316-L
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
Density: 1.57
Appearance: Grey
MDL No.: MFCD00148499