Gallium trinitrate solution (9 - 10% Ga) (w/w)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Gallium (III) nitrate
43484.30 43484.18
43484.30EA 708 EUR
43484.30 43484.18
Gallium trinitrate solution (9 - 10% Ga) (w/w)
Gallium trinitrate
Purity: 9-10% CAS No.: 13494-90-1 Molecular Formula: Ga(NO3)3 Formula Weight: 255.73 Physical Form: Liquid UN No.: UN3218 MDL No.: MFCD00011016
Formule: GaN₃O₉
MW: 255.74 g/mol
Smeltpunt: 110
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011016
CAS nummer: 13494-90-1
EINECS: 236-815-5
UN: 3218
ADR: 5.1 III