Ammonium tetrathiomolybdate ≥99.95% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
43493.14
43493.06
43493.14EA
548
EUR
43493.14
43493.06
Ammonium tetrathiomolybdate ≥99.95% (metals basis)
Ammonium tetrathiomolybdate
Purity: 99.95%
CAS No.: 15060-55-6
Molecular Formula: H8MoN2S4
Formula Weight: 260.27
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Black
MDL No.: MFCD00136013
Formule:
H₁₀MoN₂S₄ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00136013 CAS nummer: 15060-55-6 |