Polyethylene oxide, M.W. 1,000,000
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43678.22
43678.14
43678.36
43678.22EA
150
EUR
43678.22
43678.14
43678.36
Polyethylene oxide, M.W. 1,000,000
Polyethylene oxide
CAS No.: 25322-68-3
Molecular Formula: (-CH2CH2O-)n
Physical Form: Powder
Odour: Odourless
pH: 5-7
MDL No.: MFCD00147971
Formule:
(-CH₂CH₂O-)n Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00147970 CAS nummer: 25322-68-3 EINECS: 500-038-2 Merck Index: 14,7568 |