Ethyl 2-cyanoacetoacetate
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Ethyl2-acetylcyanoacetate
43792.09
43792.04
43792.09EA
146
EUR
43792.09
43792.04
Ethyl 2-cyanoacetoacetate
Ethyl-2-cyanoacetoacetate
CAS No.: 634-55-9
Molecular Formula: C2H5OOCCH(CN)COCH3
Formula Weight: 155.16
Physical Form: Liquid
UN No.: UN3276
MDL No.: MFCD00043487
Formule:
C₇H₉NO₃ MW: 155.153 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00043487 CAS nummer: 634-55-9 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...