Zinc copper composite ≥99% (metals basis) -100 mesh
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
44013.09
44013.22
44013.36
44013.09EA
26.5
EUR
44013.09
44013.22
44013.36
Zinc copper composite ≥99% (metals basis) -100 mesh
Zinc copper composite
Purity: 99%
Molecular Formula: Typically Zn 90-95, Cu 5-10 wt%
Physical Form: Alloy Precursor
UN No.: UN3089
MDL No.: MFCD03102068
Formule:
∼Zn 90-95,Cu 5-10% Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD03102068 UN: 3089 ADR: 4.1,III |