Silver plating solution ∼2,1 g/l metal content, electroless
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
44068.AK
44068.K2
44068.AKEA
38.3
EUR
44068.AK
44068.K2
Silver plating solution ∼2,1 g/l metal content, electroless
Silver plating solution
Physical Form: Liquid
Density: 1.04
Appearance: Colourless
Melting Point: -5°
Boiling Point: 103-105°
UN No.: UN3287
MDL No.: MFCD00003397
Applications: For plating on copper, brass or copper alloys
Kookpunt:
103…105 °C Smeltpunt: –5 °C Dichtheid: 1,04 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003397 UN: 3287 ADR: 6.1,II |