Semi metals, plasma standard solution, As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 µg/ml, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
44268.AE 44268.AP
44268.AEEA 194 EUR
44268.AE 44268.AP
Semi metals, plasma standard solution, As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 µg/ml, Specpure®
Standaarden Kationische standaarden Standaarden met meerdere elementen
Molecular Formula: Matrix: 20% HCl/tr. HF
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00145276