Refractory metals, plasma standard solution, Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr @ 100 µg/ml, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
44270.AE 44270.AP
44270.AEEA 239 EUR
44270.AE 44270.AP
Refractory metals, plasma standard solution, Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr @ 100 µg/ml, Specpure®
Standaarden Kationische standaarden Standaarden met meerdere elementen
UN No.: UN3264
MDL No.: MFCD00151265