Kiezelgel, granules -6+16 mesh desiccant, with indicator
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
44389.30
44389.A1
44389.A7
44389.30EA
44.5
EUR
44389.30
44389.A1
44389.A7
Kiezelgel, granules -6+16 mesh desiccant, with indicator
Kiezelgel
Re-use after being saturated with water by heating to 121 °C for approximately 8 hours
CAS No.: 7631-86-9
Physical Form: Granules
Sensitivity: Hygroscopic
Appearance: Blue to red
Odour: Odourless
pH: 6
MDL No.: MFCD00011232
Formule:
SiO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011232 CAS nummer: 63231-67-4 EINECS: 231-545-4 |