Benzyl mercaptan, polymer supported 2,0-4,0 mmol/g 2.0-4.0 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
44466.04
44466.09
44466.18
44466.04EA
49.5
EUR
44466.04
44466.09
44466.18
Benzyl mercaptan, polymer supported 2,0-4,0 mmol/g 2.0-4.0 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Benzyl mercaptan, polymer supported
Physical Form: Beads
Appearance: White to pink
MDL No.: MFCD01863867