Silicon(II) oxide, high surface area catalyst support
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Silicon monoxide, Silicon oxide
44740.22 44740.36 44740.A3
44740.22EA 45.4 EUR
44740.22 44740.36 44740.A3
Silicon(II) oxide, high surface area catalyst support
Silicon(II) oxide
Physical Form: 3.18mm (0.125in) Pellets MDL No.: MFCD00011232
Applications: Typical properties: S.A.250m2/g
Formule: SiO
MW: 44.08 g/mol
Kookpunt: 1880
Smeltpunt: 1702
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011232
CAS nummer: 10097-28-6
Merck Index: 12,08640