Rhodium 5% on activated carbon, paste C101023-5
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
44863.04 44863.09
44863.04EA 264 EUR
44863.04 44863.09
Rhodium 5% on activated carbon, paste C101023-5
Rhodium
CAS No.: 7440-16-6 Physical Form: Paste Appearance: Black MDL No.: MFCD03458392
Formule: Rh
MW: 102.9 g/mol
Smeltpunt: 1960
Dichtheid: 12.41
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011201
CAS nummer: 7440-16-6
EINECS: 231-125-0
Merck Index: 12,08353