Silver conductive ink
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
44958.14
44958.06
44958.14EA
329
EUR
44958.14
44958.06
Silver conductive ink
Silver conductive ink
Applications: This is a conducting screen printable thick film ink for membrane switch production.
CAS No.: 7440-22-4
Physical Form: Liquid
Density: 2.3
Appearance: Silver
MDL No.: MFCD01868501
Dichtheid:
2,3 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD01868501 EINECS: 231-131-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...