Monel® 400 alloy, gauze woven from wire 200 mesh woven from 0.05mm (0.0021in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45064.GT
45064.GH
45064.HB
45064.GTEA
70.7
EUR
45064.GT
45064.GH
45064.HB
Monel® 400 alloy, gauze woven from wire 200 mesh woven from 0.05mm (0.0021in)
Monel® 400 alloy
MDL No.: MFCD09910536