Monel® 400 alloy, wire, Ø 0.25 mm (0.01 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45065.TC
45065.TB
45065.TCEA
131
EUR
45065.TC
45065.TB
Monel® 400 alloy, wire, Ø 0.25 mm (0.01 in)
Monel® 400 alloy
Physical Form: ≈0.4339g/m
MDL No.: MFCD09910536